Learn More
Medchemexpress LLC 4-methylumbelliferyl-α-D-galactopyranoside | 38597-12-5 | MFCD00063278 | 99.8% | 338.31 g/mol | C16H18O8 | 50 MG
SDP
Catalog No. 5000307718
Encompass_Preferred
4-Methylumbelliferyl-α-D-galactopyranoside is a blue pro-fluorogenic substrate used to measure α-galactosidase A (GLA) activity. Enzymatic hydrolysis releases galactose and fluorescent 4-methylumbelliferone (4-MU), enabling sensitive fluorometric detection of enzyme activity in biochemical and cell-based assays. Supplied as a high-purity powder for research use only.
  • Fluorogenic substrate for α-galactosidase assays
  • Releases fluorescent 4-methylumbelliferone upon enzymatic cleavage
  • High purity (99.8%) suitable for enzymatic assays
  • Supplied as 50 mg powder
  • Recommended storage: powder at -20°C to maximize stability
Catalog No. 50-003-07718 Supplier Medchemexpress LLC Supplier No. HY11813550MG
4-Methylumbelliferyl-α-D-galactopyranoside is a blue pro-fluorogenic substrate used to measure α-galactosidase A (GLA) activity. Enzymatic hydrolysis releases galactose and fluorescent 4-methylumbelliferone (4-MU), enabling sensitive fluorometric detection of enzyme activity in biochemical and cell-based assays. Supplied as a high-purity powder for research use only.
  • Fluorogenic substrate for α-galactosidase assays
  • Releases fluorescent 4-methylumbelliferone upon enzymatic cleavage
  • High purity (99.8%) suitable for enzymatic assays
  • Supplied as 50 mg powder
  • Recommended storage: powder at -20°C to maximize stability