Learn More
Alkali Scientific 4 Methylumbelliferyl β D Galactopyranoside 6 Sulfate, Sodium Salt, 25 Mg
SDP
Catalog No. 5000661666
Encompass_Preferred
Methylumbelliferyl Galactopyranoside Sulfate is a high-purity biochemical reagent used in enzymatic assays to study galactosidase enzymes and their activity with galactose derivatives. The 25mg reagent is particularly useful in high-sensitivity tests and research focusing on the role of sulfate-modified carbohydrates. It is widely used in studies of carbohydrate metabolism and glycosidase enzyme activity in clinical and laboratory settings. This reagent’s high purity ensures reproducibility and accuracy in enzyme-linked assays and is valuable for studying the enzymatic breakdown of galactose-containing substrates in both biological and industrial applications.
Catalog No. 50-006-61666 Supplier Alkali Scientific Supplier No. BDG001CP
Methylumbelliferyl Galactopyranoside Sulfate is a high-purity biochemical reagent used in enzymatic assays to study galactosidase enzymes and their activity with galactose derivatives. The 25mg reagent is particularly useful in high-sensitivity tests and research focusing on the role of sulfate-modified carbohydrates. It is widely used in studies of carbohydrate metabolism and glycosidase enzyme activity in clinical and laboratory settings. This reagent’s high purity ensures reproducibility and accuracy in enzyme-linked assays and is valuable for studying the enzymatic breakdown of galactose-containing substrates in both biological and industrial applications.