Learn More
Alkali Scientific 4 Methylumbelliferyl α D Galactopyranoside Monohydrate [4 Methylumbelliferyl α D Galactoside], 1 G
SDP
Catalog No. 5000660213
Encompass_Preferred
Methylumbelliferyl α-D-Galactopyranoside Monohydrate (1 g) is a fluorogenic substrate that is widely used in assays to detect and quantify galactosidase activity. This reagent is critical in studying the breakdown of galactose-containing polysaccharides and glycoproteins in microbial, plant, and animal systems. Upon enzyme cleavage, it produces a fluorescent product that can be easily detected using a fluorometer, providing a sensitive method for enzyme activity measurement. The 1 g quantity is ideal for high-throughput laboratory experiments, allowing for large-scale screening and research of galactosidase enzyme activity in various biological contexts.
Catalog No. 50-006-60213 Supplier Alkali Scientific Supplier No. BEL003CP
Methylumbelliferyl α-D-Galactopyranoside Monohydrate (1 g) is a fluorogenic substrate that is widely used in assays to detect and quantify galactosidase activity. This reagent is critical in studying the breakdown of galactose-containing polysaccharides and glycoproteins in microbial, plant, and animal systems. Upon enzyme cleavage, it produces a fluorescent product that can be easily detected using a fluorometer, providing a sensitive method for enzyme activity measurement. The 1 g quantity is ideal for high-throughput laboratory experiments, allowing for large-scale screening and research of galactosidase enzyme activity in various biological contexts.