Learn More
Alkali Scientific 4 Methylumbelliferyl α D Galactopyranoside Monohydrate [4 Methylumbelliferyl α D Galactoside], 100 Milligrams
SDP
Catalog No. 5000661164
Encompass_Preferred
Methylumbelliferyl Galactopyranoside is a high-purity reagent widely used in biochemical analysis, especially in assays targeting galactosidase enzymes. This 100mg reagent is commonly employed in studies of lactose metabolism and in enzymatic assays that involve the hydrolysis of galactose-containing substrates. It is especially useful in clinical research, microbiology, and biochemistry for studying the breakdown of complex carbohydrates. This reagent is essential in enzyme-linked assays, enzyme activity screening, and metabolic research focusing on galactose or lactose metabolism, providing valuable data on enzyme kinetics and activity.
Catalog No. 50-006-61164 Supplier Alkali Scientific Supplier No. BEL004CP
Methylumbelliferyl Galactopyranoside is a high-purity reagent widely used in biochemical analysis, especially in assays targeting galactosidase enzymes. This 100mg reagent is commonly employed in studies of lactose metabolism and in enzymatic assays that involve the hydrolysis of galactose-containing substrates. It is especially useful in clinical research, microbiology, and biochemistry for studying the breakdown of complex carbohydrates. This reagent is essential in enzyme-linked assays, enzyme activity screening, and metabolic research focusing on galactose or lactose metabolism, providing valuable data on enzyme kinetics and activity.