Learn More
Alkali Scientific 4 Methylumbelliferyl α D Galactopyranoside Monohydrate (4 Methylumbelliferyl α D Galactoside), 50 Mg
SDP
Catalog No. 5000660672
Encompass_Preferred
Methylumbelliferyl α-D-Galactopyranoside Monohydrate (50 mg) is a fluorogenic substrate used for the measurement of galactosidase activity. This substrate is widely used in biochemical assays and research to detect the enzymatic hydrolysis of galactosidases in microbial and mammalian systems. The fluorogenic nature of this compound allows for easy detection and quantification of enzyme activity through fluorescence emission. It is particularly useful in studying glycosidases, and this 50 mg quantity is suitable for various laboratory applications including high-throughput assays or routine screening of galactosidase activity in samples.
Catalog No. 50-006-60672 Supplier Alkali Scientific Supplier No. BEL001CP
Methylumbelliferyl α-D-Galactopyranoside Monohydrate (50 mg) is a fluorogenic substrate used for the measurement of galactosidase activity. This substrate is widely used in biochemical assays and research to detect the enzymatic hydrolysis of galactosidases in microbial and mammalian systems. The fluorogenic nature of this compound allows for easy detection and quantification of enzyme activity through fluorescence emission. It is particularly useful in studying glycosidases, and this 50 mg quantity is suitable for various laboratory applications including high-throughput assays or routine screening of galactosidase activity in samples.