Learn More
Bioworld 4-Methylumbelliferyl-á-D-Glucopyranoside, 250 mg
Catalog No. 50198772
Encompass_Preferred
4-Methylumbelliferyl-α-D-glucopyranoside[4-Methylumbelliferyl-α-D-Glucoside]C16H18O8MW 338.3Fluorescent substrate for -α-D-Glucosidase.Purity: >98%. 4-Methylumbelliferone <50ppm.Soluble in 2% acetone/water 1:1.CAS 17833-43-1This product is sold for laboratory research use only.
Catalog No. 50-198-772 Supplier Bioworld Supplier No. 215300082
4-Methylumbelliferyl-α-D-glucopyranoside[4-Methylumbelliferyl-α-D-Glucoside]C16H18O8MW 338.3Fluorescent substrate for -α-D-Glucosidase.Purity: >98%. 4-Methylumbelliferone <50ppm.Soluble in 2% acetone/water 1:1.CAS 17833-43-1This product is sold for laboratory research use only.