Learn More
Bioworld 4-Methylumbelliferyl-B-D-Glucopyranoside, 250 mg
Catalog No. 50198791
Encompass_Preferred
4-Methylumbelliferyl-β-D-glucopyranoside C18H18O8MW 338.3 Fluorescent substrate for β-D-Glucosidase.Purity: >98%. 4-Methylumbelliferone <50ppm.Soluble in 2% acetone/water 1:1.CAS 18997-57-4. This product is sold for laboratory research use only.
Catalog No. 50-198-791 Supplier Bioworld Supplier No. 215300181
4-Methylumbelliferyl-β-D-glucopyranoside C18H18O8MW 338.3 Fluorescent substrate for β-D-Glucosidase.Purity: >98%. 4-Methylumbelliferone <50ppm.Soluble in 2% acetone/water 1:1.CAS 18997-57-4. This product is sold for laboratory research use only.