Learn More
Medchemexpress LLC 4-Methylumbelliferyl-α-D-Glucopyranoside | 17833-43-1 | >99.9% | 338.31 | 50 MG
SDP
Catalog No. 5000350979
Encompass_Preferred
4-Methylumbelliferyl-α-D-Glucopyranoside (4-Methylumbelliferyl-α-D-Glucose) is a fluorescent substrate for α-glucosidase. It releases the fluorescent moiety 4-methylumbelliferyl (4-MU) upon cleavage, which exhibits pH-dependent fluorescence excitation. This compound can be utilized as a biomarker for Fabry and Pompe diseases to quantify α-glucosidase activity in infant blood spot samples.
  • Fluorescent substrate for alpha-glucosidase
  • Releases 4-methylumbelliferyl (4-MU) upon cleavage
  • 4-MU has pH-dependent fluorescence excitation activity
  • Excitation wavelengths are 320 nm at low pH (1.97-6.72) and 360 nm at high pH (7.12-10.3)
  • Emission wavelength increases with decreasing pH, ranging from 445-455 nm
  • Useful as a biomarker for Fabry and Pompe diseases
  • Quantifies alpha-glucosidase activity in infant blood spot samples
Catalog No. 50-003-50979 Supplier Medchemexpress LLC Supplier No. HY13787350MG
4-Methylumbelliferyl-α-D-Glucopyranoside (4-Methylumbelliferyl-α-D-Glucose) is a fluorescent substrate for α-glucosidase. It releases the fluorescent moiety 4-methylumbelliferyl (4-MU) upon cleavage, which exhibits pH-dependent fluorescence excitation. This compound can be utilized as a biomarker for Fabry and Pompe diseases to quantify α-glucosidase activity in infant blood spot samples.
  • Fluorescent substrate for alpha-glucosidase
  • Releases 4-methylumbelliferyl (4-MU) upon cleavage
  • 4-MU has pH-dependent fluorescence excitation activity
  • Excitation wavelengths are 320 nm at low pH (1.97-6.72) and 360 nm at high pH (7.12-10.3)
  • Emission wavelength increases with decreasing pH, ranging from 445-455 nm
  • Useful as a biomarker for Fabry and Pompe diseases
  • Quantifies alpha-glucosidase activity in infant blood spot samples