Learn More
Medchemexpress LLC 4-Methylumbelliferyl-α-D-Glucopyranoside | 17833-43-1 | 99.99% | 338.31 | 25 MG
SDP
Catalog No. 5000347808
Encompass_Preferred
4-Methylumbelliferyl-α-D-Glucopyranoside is a fluorescent substrate for α-glucosidase, which releases the fluorescent moiety 4-methylumbelliferyl (4-MU) upon cleavage. 4-MU exhibits pH-dependent fluorescence excitation activity, with excitation wavelengths of 320 nm at low pH (1.97-6.72) and 360 nm at high pH (7.12-10.3). The emission wavelength of 4-Methylumbelliferyl-α-D-Glucopyranoside increases with decreasing pH, ranging from 445-455 nm. This compound can be utilized as a biomarker for Fabry and Pompe diseases to quantify α-glucosidase activity in infant blood spot samples.
  • Fluorescent substrate for α-glucosidase.
  • Releases fluorescent moiety 4-methylumbelliferyl (4-MU) upon cleavage.
  • 4-MU has pH-dependent fluorescence excitation activity.
  • Can be used as a biomarker for Fabry and Pompe diseases.
  • Quantifies α-glucosidase activity in infant blood spot samples.
Catalog No. 50-003-47808 Supplier Medchemexpress LLC Supplier No. 25MGHY137873
4-Methylumbelliferyl-α-D-Glucopyranoside is a fluorescent substrate for α-glucosidase, which releases the fluorescent moiety 4-methylumbelliferyl (4-MU) upon cleavage. 4-MU exhibits pH-dependent fluorescence excitation activity, with excitation wavelengths of 320 nm at low pH (1.97-6.72) and 360 nm at high pH (7.12-10.3). The emission wavelength of 4-Methylumbelliferyl-α-D-Glucopyranoside increases with decreasing pH, ranging from 445-455 nm. This compound can be utilized as a biomarker for Fabry and Pompe diseases to quantify α-glucosidase activity in infant blood spot samples.
  • Fluorescent substrate for α-glucosidase.
  • Releases fluorescent moiety 4-methylumbelliferyl (4-MU) upon cleavage.
  • 4-MU has pH-dependent fluorescence excitation activity.
  • Can be used as a biomarker for Fabry and Pompe diseases.
  • Quantifies α-glucosidase activity in infant blood spot samples.