Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-methylumbelliferyl α-D-glucopyranoside | 17833-43-1 | MFCD00067661 | 100.0% | 338.31 g·mol⁻¹ | C16H18O8 | 5 MG
SDP

Catalog No. 5000222673
Encompass_Preferred

4-Methylumbelliferyl α-D-glucopyranoside is a fluorogenic substrate for α-glucosidase that releases 4-methylumbelliferone upon enzymatic hydrolysis. The liberated fluorophore enables sensitive, quantitative measurement of GAA activity in biochemical assays and newborn screening workflows for lysosomal storage disorders. The reagent is provided in small research quantities and requires controlled storage to preserve stability.

  • Fluorogenic substrate for α-glucosidase assays.
  • Enzymatic cleavage yields fluorescent 4-methylumbelliferone for quantitation.
  • Excitation ~360 nm, emission ~445-455 nm (read at ~450 nm).
  • Applicable to newborn screening and research on lysosomal storage disorders.
  • Supplied in small research-scale quantities; follow storage recommendations.
  • Storage: store at 4°C under inert gas; in solution, store at -80°C or -20°C as specified.

Catalog No. 50-002-22673 Supplier Medchemexpress LLC Supplier No. HY1378735MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Methylumbelliferyl α-D-glucopyranoside is a fluorogenic substrate for α-glucosidase that releases 4-methylumbelliferone upon enzymatic hydrolysis. The liberated fluorophore enables sensitive, quantitative measurement of GAA activity in biochemical assays and newborn screening workflows for lysosomal storage disorders. The reagent is provided in small research quantities and requires controlled storage to preserve stability.

  • Fluorogenic substrate for α-glucosidase assays.
  • Enzymatic cleavage yields fluorescent 4-methylumbelliferone for quantitation.
  • Excitation ~360 nm, emission ~445-455 nm (read at ~450 nm).
  • Applicable to newborn screening and research on lysosomal storage disorders.
  • Supplied in small research-scale quantities; follow storage recommendations.
  • Storage: store at 4°C under inert gas; in solution, store at -80°C or -20°C as specified.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.