Learn More
Medchemexpress LLC 4-methylumbelliferyl β-d-glucopyranoside | 18997-57-4 | MFCD00063694 | 99.9% | 338.31 g/mol | C16H18O8 | 1G
SDP
Catalog No. 5000238886
Encompass_Preferred
4-Methylumbelliferyl β-D-Glucopyranoside is a fluorogenic β-D-glucoside substrate used to measure β-glucosidase activity. Enzymatic cleavage releases 4-methylumbelliferone (4-MU), a highly fluorescent reporter (emission 445-454 nm); excitation maxima are pH-dependent (330, 370, 385 nm at pH 4.6, 7.4, 10.4). The product is supplied as a solid and as a 10 mM solution in DMSO for convenient assay setup.
  • Fluorogenic substrate for β-glucosidase assays.
  • Releases 4-methylumbelliferone for sensitive fluorescence detection.
  • High reported purity (99.87%).
  • Available as solid and 10 mM solution in DMSO.
  • Suitable for biochemical and cell-based enzyme activity assays.
  • Molecular weight 338.31 g/mol; formula C16H18O8.
  • Pack sizes suitable for assay workflows, including 1 g solid options.
Catalog No. 50-002-38886 Supplier Medchemexpress LLC Supplier No. HY1236331G
4-Methylumbelliferyl β-D-Glucopyranoside is a fluorogenic β-D-glucoside substrate used to measure β-glucosidase activity. Enzymatic cleavage releases 4-methylumbelliferone (4-MU), a highly fluorescent reporter (emission 445-454 nm); excitation maxima are pH-dependent (330, 370, 385 nm at pH 4.6, 7.4, 10.4). The product is supplied as a solid and as a 10 mM solution in DMSO for convenient assay setup.
  • Fluorogenic substrate for β-glucosidase assays.
  • Releases 4-methylumbelliferone for sensitive fluorescence detection.
  • High reported purity (99.87%).
  • Available as solid and 10 mM solution in DMSO.
  • Suitable for biochemical and cell-based enzyme activity assays.
  • Molecular weight 338.31 g/mol; formula C16H18O8.
  • Pack sizes suitable for assay workflows, including 1 g solid options.