Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-methylumbelliferyl β-D-glucopyranoside | 18997-57-4 | MFCD00063694 | >98.0% | 338.31 g/mol | C16H18O8 | 500MG
SDP

Catalog No. 5000236707
Encompass_Preferred

4-Methylumbelliferyl β-D-glucopyranoside is a fluorogenic β-D-glucoside substrate for measuring β-glucosidase activity. Upon enzymatic cleavage it releases 4-methylumbelliferone (4-MU), a highly fluorescent product useful for quantitative enzymatic assays and high-throughput screening. The excitation maxima of 4-MU are pH-dependent, and the compound is provided as a white to off-white powder suitable for laboratory use.

  • Fluorogenic substrate for β-glucosidase assays.
  • Releases 4-methylumbelliferone with emission ~445-454 nm.
  • Excitation maxima depend on pH (≈330, 370, 385 nm).
  • High purity (≥98.0% by HPLC) for reliable results.
  • Available in multiple pack sizes, including 500 MG.
  • Molecular formula C16H18O8; molecular weight 338.31 g/mol.

Catalog No. 50-002-36707 Supplier Medchemexpress LLC Supplier No. HY123633500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Methylumbelliferyl β-D-glucopyranoside is a fluorogenic β-D-glucoside substrate for measuring β-glucosidase activity. Upon enzymatic cleavage it releases 4-methylumbelliferone (4-MU), a highly fluorescent product useful for quantitative enzymatic assays and high-throughput screening. The excitation maxima of 4-MU are pH-dependent, and the compound is provided as a white to off-white powder suitable for laboratory use.

  • Fluorogenic substrate for β-glucosidase assays.
  • Releases 4-methylumbelliferone with emission ~445-454 nm.
  • Excitation maxima depend on pH (≈330, 370, 385 nm).
  • High purity (≥98.0% by HPLC) for reliable results.
  • Available in multiple pack sizes, including 500 MG.
  • Molecular formula C16H18O8; molecular weight 338.31 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.