Learn More
Medchemexpress LLC 4-Methylumbelliferyl-β-D-glucuronide hydrate | 881005-91-0 | 99.9% | 1 ML
SDP
Catalog No. 5000409607
Encompass_Preferred
4-Methylumbelliferyl-β-D-glucuronide hydrate is a fluorogenic substrate with an excitation wavelength of 362 nm and an emission wavelength of 445 nm. This compound is used in various enzymatic reactions and has applications in detecting β-glucuronidase activity and quantifying *Escherichia coli*.
  • Fluorogenic substrate with λex=362 nm and λem=445 nm
  • Detects activity of β-glucuronidase
  • Quantifies *Escherichia coli*
  • Releases 4-methylumbelliferone in enzymatic reactions
  • Suitable for kinase assays to measure GLUase activity
Catalog No. 50-004-09607 Supplier Medchemexpress LLC Supplier No. HYD0935A10MM/1ML
4-Methylumbelliferyl-β-D-glucuronide hydrate is a fluorogenic substrate with an excitation wavelength of 362 nm and an emission wavelength of 445 nm. This compound is used in various enzymatic reactions and has applications in detecting β-glucuronidase activity and quantifying *Escherichia coli*.
  • Fluorogenic substrate with λex=362 nm and λem=445 nm
  • Detects activity of β-glucuronidase
  • Quantifies *Escherichia coli*
  • Releases 4-methylumbelliferone in enzymatic reactions
  • Suitable for kinase assays to measure GLUase activity