Learn More
Medchemexpress LLC 4-Methylumbelliferyl-β-D-glucuronide hydrate | 881005-91-0 | 99.92% | 100 MG
SDP
Catalog No. 5000410082
Encompass_Preferred
4-Methylumbelliferyl-β-D-glucuronide hydrate is a fluorogenic substrate (λex=362 nm, λem=445 nm). It has potential applications in detecting the activity of β-glucuronidase and the number of Escherichia coli. It is highly soluble in DMSO (150 mg/mL) and moderately soluble in H2O (5.2 mg/mL), requiring sonication for dissolution.
  • Fluorogenic substrate with specific excitation and emission wavelengths
  • Detects activity of β-glucuronidase
  • Used for detecting Escherichia coli
  • High solubility in DMSO
  • Requires sonication for dissolution in water
Catalog No. 50-004-10082 Supplier Medchemexpress LLC Supplier No. HYD0935A100MG
4-Methylumbelliferyl-β-D-glucuronide hydrate is a fluorogenic substrate (λex=362 nm, λem=445 nm). It has potential applications in detecting the activity of β-glucuronidase and the number of Escherichia coli. It is highly soluble in DMSO (150 mg/mL) and moderately soluble in H2O (5.2 mg/mL), requiring sonication for dissolution.
  • Fluorogenic substrate with specific excitation and emission wavelengths
  • Detects activity of β-glucuronidase
  • Used for detecting Escherichia coli
  • High solubility in DMSO
  • Requires sonication for dissolution in water