Learn More
Bioworld 4-Methylumbelliferyl-A-D-mannopyranoside, 25 mg
Catalog No. 50198774
Encompass_Preferred
C16H18O8MW 338.3 Fluorescent substrate for α-D-Mannosidase. * Purity: >98% * 4-Methylumbelliferone ,50ppm * Soluble in 2% acetone/water 1:1 * Cas No. 28541-83-5 This product is sold for laboratory research use only.
Catalog No. 50-198-774 Supplier Bioworld Supplier No. 215300091
C16H18O8MW 338.3 Fluorescent substrate for α-D-Mannosidase. * Purity: >98% * 4-Methylumbelliferone ,50ppm * Soluble in 2% acetone/water 1:1 * Cas No. 28541-83-5 This product is sold for laboratory research use only.