Learn More
Alkali Scientific 4 Methylumbelliferyl ? D mannopyranoside, 100 Mg
SDP
Catalog No. 5000660601
Encompass_Preferred
4 Methylumbelliferyl ? D Mannopyranoside, available in a 100 mg pack, is a specialized reagent used to detect mannanase activity in biochemical and microbiological assays. When hydrolyzed by mannanase enzymes, this compound generates a fluorescent signal that is easily quantified. This reagent is essential in research on carbohydrate metabolism, particularly the breakdown of mannans in plant biomass. The 100 mg pack is suitable for repeated use in laboratory testing, making it ideal for high-throughput analysis. It is commonly employed in studies on enzyme kinetics, biofuel production, and other industrial applications requiring polysaccharide degradation.
Catalog No. 50-006-60601 Supplier Alkali Scientific Supplier No. YLD002CP
4 Methylumbelliferyl ? D Mannopyranoside, available in a 100 mg pack, is a specialized reagent used to detect mannanase activity in biochemical and microbiological assays. When hydrolyzed by mannanase enzymes, this compound generates a fluorescent signal that is easily quantified. This reagent is essential in research on carbohydrate metabolism, particularly the breakdown of mannans in plant biomass. The 100 mg pack is suitable for repeated use in laboratory testing, making it ideal for high-throughput analysis. It is commonly employed in studies on enzyme kinetics, biofuel production, and other industrial applications requiring polysaccharide degradation.