Learn More
Alkali Scientific 4 Methylumbelliferyl ? D mannopyranoside, 25 Mg
SDP
Catalog No. 5000660584
Encompass_Preferred
4 Methylumbelliferyl ? D Mannopyranoside is a high-purity reagent designed for the detection of mannanase activity in various biochemical assays. Upon hydrolysis by mannanase enzymes, the compound produces a fluorescent product that can be quantitatively measured. This makes it a vital tool for studying the enzymatic breakdown of mannans, a type of polysaccharide found in plant cell walls. The 25 mg pack ensures sufficient material for regular testing in both research and industrial applications. It is widely used in studies related to polysaccharide degradation, including applications in biofuel production and food processing.
Catalog No. 50-006-60584 Supplier Alkali Scientific Supplier No. YLD001CP
4 Methylumbelliferyl ? D Mannopyranoside is a high-purity reagent designed for the detection of mannanase activity in various biochemical assays. Upon hydrolysis by mannanase enzymes, the compound produces a fluorescent product that can be quantitatively measured. This makes it a vital tool for studying the enzymatic breakdown of mannans, a type of polysaccharide found in plant cell walls. The 25 mg pack ensures sufficient material for regular testing in both research and industrial applications. It is widely used in studies related to polysaccharide degradation, including applications in biofuel production and food processing.