Learn More
Medchemexpress LLC 4-Methylumbelliferyl-α-d-mannoside | 28541-83-5 | 99.3% | 338.31 | 5 MG
SDP
Catalog No. 5000453145
Encompass_Preferred
4-Methylumbelliferyl-α-d-mannoside is a fluorescent substrate for α-D-mannosidase. For research use only.
  • Fluorescent substrate for α-D-mannosidase.
  • For research use only.
Catalog No. 50-004-53145 Supplier Medchemexpress LLC Supplier No. HYW0091225MG
4-Methylumbelliferyl-α-d-mannoside is a fluorescent substrate for α-D-mannosidase. For research use only.
  • Fluorescent substrate for α-D-mannosidase.
  • For research use only.