Learn More
Bioworld 4-Methylumbelliferyl-B-D-xylopyranoside, 1 g
Catalog No. 50198798
Encompass_Preferred
C15H16O7 MW 308.3 Fluorescent substrate for β-D-Xylosidase. * Purity: >98% * 4-Methylumbelliferone <50ppm * Soluble in 0.3% pyridine * Cas No. 6734-33-4 . This product is sold for laboratory research use only.
Catalog No. 50-198-798 Supplier Bioworld Supplier No. 215300202
C15H16O7 MW 308.3 Fluorescent substrate for β-D-Xylosidase. * Purity: >98% * 4-Methylumbelliferone <50ppm * Soluble in 0.3% pyridine * Cas No. 6734-33-4 . This product is sold for laboratory research use only.