Learn More
Alkali Scientific 4 Methylumbelliferyl β D Xylopyranoside, 1 G
SDP
Catalog No. 5000660772
Encompass_Preferred
4 Methylumbelliferyl β D Xylopyranoside, available in a 1g pack, is a highly sensitive reagent used for the detection of xylanase activity in biochemical assays. This substrate, when hydrolyzed by xylanase enzymes, produces a fluorescent signal, making it ideal for high-throughput enzymatic assays. This reagent is commonly used in microbiology and biotechnology research, particularly in studies on lignocellulosic biomass degradation and the enzymatic breakdown of plant cell wall components. The 1g pack ensures ample supply for extended research needs, providing excellent value for labs focused on enzyme activity profiling and industrial applications of xylanase.
Catalog No. 50-006-60772 Supplier Alkali Scientific Supplier No. BDX002CP
Add to Cart
Add to Cart
4 Methylumbelliferyl β D Xylopyranoside, available in a 1g pack, is a highly sensitive reagent used for the detection of xylanase activity in biochemical assays. This substrate, when hydrolyzed by xylanase enzymes, produces a fluorescent signal, making it ideal for high-throughput enzymatic assays. This reagent is commonly used in microbiology and biotechnology research, particularly in studies on lignocellulosic biomass degradation and the enzymatic breakdown of plant cell wall components. The 1g pack ensures ample supply for extended research needs, providing excellent value for labs focused on enzyme activity profiling and industrial applications of xylanase.