Learn More
AdipoGen 4-Methylumbelliferyl heptanoat
SDP
Catalog No. 502560352
Encompass_Preferred
Chemical. CAS 18319-92-1. Formula C17H20O4. MW 288.34. Synthetic. Fluorogenic substrate for lipases.
Catalog No. 50-256-0352 Supplier AdipoGen Supplier No. CDXO0007M500
Chemical. CAS 18319-92-1. Formula C17H20O4. MW 288.34. Synthetic. Fluorogenic substrate for lipases.