Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-methylumbelliferyl α-L-arabinofuranoside | 77471-44-4 | MFCD00057308 | 99.9% | 308.28 | C15H16O7 | 5 MG
SDP

Catalog No. 5000223403
Encompass_Preferred

4-Methylumbelliferyl α-L-arabinofuranoside is a fluorogenic glycosidase substrate used to measure α-L-arabinofuranosidase activity. Enzymatic hydrolysis releases fluorescent 4-methylumbelliferone, enabling sensitive fluorometric assays in biochemical and enzymology workflows. For research use only.

  • Fluorogenic substrate for α-L-arabinofuranosidase assays.
  • Produces 4-methylumbelliferone for sensitive fluorometric detection.
  • High purity (99.93%) to minimize background signal.
  • Molecular weight 308.28 and formula C15H16O7.
  • Soluble in DMSO at ≥ 50 mg/mL.
  • White to off-white solid with recommended cold storage for stability.
  • Supplied in small research pack sizes suitable for assay development.

Catalog No. 50-002-23403 Supplier Medchemexpress LLC Supplier No. HY1374905MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Methylumbelliferyl α-L-arabinofuranoside is a fluorogenic glycosidase substrate used to measure α-L-arabinofuranosidase activity. Enzymatic hydrolysis releases fluorescent 4-methylumbelliferone, enabling sensitive fluorometric assays in biochemical and enzymology workflows. For research use only.

  • Fluorogenic substrate for α-L-arabinofuranosidase assays.
  • Produces 4-methylumbelliferone for sensitive fluorometric detection.
  • High purity (99.93%) to minimize background signal.
  • Molecular weight 308.28 and formula C15H16O7.
  • Soluble in DMSO at ≥ 50 mg/mL.
  • White to off-white solid with recommended cold storage for stability.
  • Supplied in small research pack sizes suitable for assay development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.