Learn More
Bioworld 4-Methylumbelliferyl-A-L-Iduronide, 2 mg
Supplier:  Bioworld 215300121
Encompass_Preferred
Substrate for alpha-L-IduronidaseSpecification: * Slightly soluble in water This product is sold for laboratory research use onlyReference:Use of 4-methylumbelliferryl-alpha-L-iduronide and 4-trifluoromethylumbelliferryl-alpha-L-iduronide for detecting alpha-L-iduronidase deficiencies in human tissue and for rapid prenatal diagnosis of Hurler disease. Vopr Med Khim. 1991 Jan-Feb; 37 (1):74-7.
Catalog No. 50-198-778