Learn More
Medchemexpress LLC 4-methylumbelliferyl-α-L-iduronide 2-sulfate sodium | 2181774-46-7 | >=98.0% | 476.32 | C16H14Na2O12S | 1 MG
SDP
Catalog No. 5000284172
Encompass_Preferred
4-Methylumbelliferyl-α-L-iduronide 2-sulfate sodium is a fluorogenic enzymatic substrate used to measure iduronate-2-sulfatase (I2S) activity. Enzymatic hydrolysis releases the fluorescent reporter 4-methylumbelliferone (4-MU), enabling quantitative fluorescence-based assays used in lysosomal storage disease research, including screening and diagnostic applications for disorders such as Hurler syndrome. Supplied as a sodium salt powder.
  • Fluorogenic substrate for iduronate-2-sulfatase (I2S) assays.
  • Releases 4-methylumbelliferone (4-MU) upon hydrolysis for fluorescence detection.
  • Suitable for quantitative enzyme activity and diagnostic research applications.
  • Provided as a sodium salt powder for storage and handling convenience.
  • CAS number 2181774-46-7; molecular weight 476.32; formula C16H14Na2O12S.
Catalog No. 50-002-84172 Supplier Medchemexpress LLC Supplier No. HYW6542721MG
4-Methylumbelliferyl-α-L-iduronide 2-sulfate sodium is a fluorogenic enzymatic substrate used to measure iduronate-2-sulfatase (I2S) activity. Enzymatic hydrolysis releases the fluorescent reporter 4-methylumbelliferone (4-MU), enabling quantitative fluorescence-based assays used in lysosomal storage disease research, including screening and diagnostic applications for disorders such as Hurler syndrome. Supplied as a sodium salt powder.
  • Fluorogenic substrate for iduronate-2-sulfatase (I2S) assays.
  • Releases 4-methylumbelliferone (4-MU) upon hydrolysis for fluorescence detection.
  • Suitable for quantitative enzyme activity and diagnostic research applications.
  • Provided as a sodium salt powder for storage and handling convenience.
  • CAS number 2181774-46-7; molecular weight 476.32; formula C16H14Na2O12S.