Learn More
Medchemexpress LLC 4-methylumbelliferyl α-L-iduronide | 66966-09-4 | MFCD00135894 | 99.0% | 352.29 g/mol | C16H16O9 | 1 MG
SDP
Catalog No. 5000283636
Encompass_Preferred
4-Methylumbelliferyl α-L-iduronide is a fluorogenic biochemical reagent used as a substrate to measure α-L-iduronidase activity. Enzymatic cleavage releases 4-methylumbelliferone, producing a fluorescent signal for sensitive fluorometric assays in lysosomal enzyme studies and biochemical assay development.
  • Fluorogenic substrate for α-L-iduronidase assays.
  • Enables sensitive fluorometric detection via 4-methylumbelliferone release.
  • Suitable for enzyme kinetics and biochemical assay development.
  • Available in small research quantities (μg-mg scale).
  • Store protected from light at -20°C; in solvent, -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-83636 Supplier Medchemexpress LLC Supplier No. HYW1277081MG
4-Methylumbelliferyl α-L-iduronide is a fluorogenic biochemical reagent used as a substrate to measure α-L-iduronidase activity. Enzymatic cleavage releases 4-methylumbelliferone, producing a fluorescent signal for sensitive fluorometric assays in lysosomal enzyme studies and biochemical assay development.
  • Fluorogenic substrate for α-L-iduronidase assays.
  • Enables sensitive fluorometric detection via 4-methylumbelliferone release.
  • Suitable for enzyme kinetics and biochemical assay development.
  • Available in small research quantities (μg-mg scale).
  • Store protected from light at -20°C; in solvent, -80°C (6 months) or -20°C (1 month).