Learn More
Medchemexpress LLC 4-methylumbelliferyl α-L-iduronide | 66966-09-4 | MFCD00135894 | ≥99.0% | 352.29 g/mol | C16H16O9 | 500 UG
SDP
Catalog No. 5000282819
Encompass_Preferred
4-Methylumbelliferyl α-L-iduronide is a fluorogenic reagent used as a sensitive substrate for α-L-iduronidase and related lysosomal enzyme assays. Enzymatic hydrolysis releases 4-methylumbelliferone, permitting fluorescence-based detection and quantitation in biochemical and cell-based assays.
  • Fluorogenic substrate for α-L-iduronidase assays.
  • Enables sensitive fluorescence detection and quantitation.
  • High purity (≥99.0%) suitable for biochemical applications.
  • Available in small research quantities (for example, 500 μg) for assay development.
  • Stable solid form for convenient storage and handling.
Catalog No. 50-002-82819 Supplier Medchemexpress LLC Supplier No. HYW127708500UG
4-Methylumbelliferyl α-L-iduronide is a fluorogenic reagent used as a sensitive substrate for α-L-iduronidase and related lysosomal enzyme assays. Enzymatic hydrolysis releases 4-methylumbelliferone, permitting fluorescence-based detection and quantitation in biochemical and cell-based assays.
  • Fluorogenic substrate for α-L-iduronidase assays.
  • Enables sensitive fluorescence detection and quantitation.
  • High purity (≥99.0%) suitable for biochemical applications.
  • Available in small research quantities (for example, 500 μg) for assay development.
  • Stable solid form for convenient storage and handling.