Learn More
AdipoGen 4-Methylumbelliferyl nonanoate
SDP
Catalog No. 502560472
Encompass_Preferred
Chemical. CAS 18319-93-2. Formula C19H24O4. MW 316.39. Synthetic. Non-fluorescent lipase substrate conjugated with 4-methylumbelliferone. After lipolytic hydrolysed the highly fluorescent 4-methylumbelliferone is released and emitts strongly at 485 nm blue fluorescence ex = 366 nm.
Catalog No. 50-256-0472 Supplier AdipoGen Supplier No. CDXM0219G005
Chemical. CAS 18319-93-2. Formula C19H24O4. MW 316.39. Synthetic. Non-fluorescent lipase substrate conjugated with 4-methylumbelliferone. After lipolytic hydrolysed the highly fluorescent 4-methylumbelliferone is released and emitts strongly at 485 nm blue fluorescence ex = 366 nm.