Learn More
AdipoGen 4-Methylumbelliferyl octanoate
SDP
Catalog No. 502560474
Encompass_Preferred
Chemical. CAS 20671-66-3. Formula C18H22O4. MW 302.36. Synthetic. Fluorogenic substrate for caprylate esterase used as tool to identify salmonella in clinical labs. MUCAP consists of a flexible eight carbon atom ester conjugated with 4-methylumbelliferone. The non-fluorescent MUCAP substrate interacts with the salmonella C8 esterase in a specific manner leading to the release of highly fluorescent 4-methylumbelliferone that emitts strongly at 485 nm blue fluorescence ex = 366 nm.
Catalog No. 50-256-0474 Supplier AdipoGen Supplier No. CDXM0218G005
Chemical. CAS 20671-66-3. Formula C18H22O4. MW 302.36. Synthetic. Fluorogenic substrate for caprylate esterase used as tool to identify salmonella in clinical labs. MUCAP consists of a flexible eight carbon atom ester conjugated with 4-methylumbelliferone. The non-fluorescent MUCAP substrate interacts with the salmonella C8 esterase in a specific manner leading to the release of highly fluorescent 4-methylumbelliferone that emitts strongly at 485 nm blue fluorescence ex = 366 nm.