Learn More
AdipoGen 4-Methylumbelliferyl oleate,Chemical. CAS: 18323-58-5
SDP
Supplier:  AdipoGen CDXM0086M025
Encompass_Preferred
4-Methylumbelliferyl oleate, AdipoGen Life Sciences. Chemical. CAS: 18323-58-5. Formula: C28H40O4. MW: 440.61. Synthetic. Fluorogenic substrate for lipases, which can be used in aqueous solutions as well as several organic solvents.
Catalog No. 50-595-940