Learn More
AdipoGen 4-Methylumbelliferyl oleate
SDP
Catalog No. 502560530
Encompass_Preferred
Chemical. CAS 18323-58-5. Formula C28H40O4. MW 440.61. Synthetic. Fluorogenic substrate for lipases, which can be used in aqueous solutions as well as several organic solvents.
Catalog No. 50-256-0530 Supplier AdipoGen Supplier No. CDXM0086GG25
Chemical. CAS 18323-58-5. Formula C28H40O4. MW 440.61. Synthetic. Fluorogenic substrate for lipases, which can be used in aqueous solutions as well as several organic solvents.