Learn More
AdipoGen 4-Methylumbelliferyl palmitate
SDP
Catalog No. 502560266
Encompass_Preferred
Chemical. CAS 17695-48-6. Formula C26H38O4. Molecular Weight 414.59. 4-Methylumbelliferyl palmitate 4-MUP is a fluorogenic substrate for lysosomal acid lypase LAL. 4-MUP is cleaved by LAL or by other acid lipases to release the fluorescent moiety 4-MU. Recent advances allow the assessment of LAL activity in very small blood volumes using 4-MUP. Cholesterol ester storage disease and Wolman disease are recessive autosomal disorders caused by a deficiency in lysosomal acid lipase LAL, also known as cholesteryl ester hydrolase. Spectral data 4-MU fluorescence is pH-dependent. lambdaex 320nm pH 1.97-6.72 and 360nm pH 7.12-10.3 lambdaem 445-455nM increasing as pH decreases.
Catalog No. 50-256-0266 Supplier AdipoGen Supplier No. CDXP0038M250
Chemical. CAS 17695-48-6. Formula C26H38O4. Molecular Weight 414.59. 4-Methylumbelliferyl palmitate 4-MUP is a fluorogenic substrate for lysosomal acid lypase LAL. 4-MUP is cleaved by LAL or by other acid lipases to release the fluorescent moiety 4-MU. Recent advances allow the assessment of LAL activity in very small blood volumes using 4-MUP. Cholesterol ester storage disease and Wolman disease are recessive autosomal disorders caused by a deficiency in lysosomal acid lipase LAL, also known as cholesteryl ester hydrolase. Spectral data 4-MU fluorescence is pH-dependent. lambdaex 320nm pH 1.97-6.72 and 360nm pH 7.12-10.3 lambdaem 445-455nM increasing as pH decreases.