Learn More
Medchemexpress LLC 4-Methylumbelliferyl palmitate | 17695-48-6 | 99.2% | 414.59 | 10 MG
SDP
Catalog No. 5000366278
Encompass_Preferred
4-Methylumbelliferyl palmitate is an excellent fluorophore for measuring acid lipase in human leukocytes. Acidity and solvent have important influence on its fluorescence. It exists mainly as neutral molecular form which can be produced strong fluorescence at 445 nm in near neutral aqueous solutions, and exists mainly as anion form which can be produced stronger fluorescence at 445 nm in weak alkaline solutions.
  • Excellent fluorophore for measuring acid lipase in human leukocytes.
  • Fluorescence is influenced by acidity and solvent.
  • Produces strong fluorescence at 445 nm in near neutral aqueous solutions.
  • Produces stronger fluorescence at 445 nm in weak alkaline solutions.
Catalog No. 50-003-66278 Supplier Medchemexpress LLC Supplier No. HYW01094710MG
4-Methylumbelliferyl palmitate is an excellent fluorophore for measuring acid lipase in human leukocytes. Acidity and solvent have important influence on its fluorescence. It exists mainly as neutral molecular form which can be produced strong fluorescence at 445 nm in near neutral aqueous solutions, and exists mainly as anion form which can be produced stronger fluorescence at 445 nm in weak alkaline solutions.
  • Excellent fluorophore for measuring acid lipase in human leukocytes.
  • Fluorescence is influenced by acidity and solvent.
  • Produces strong fluorescence at 445 nm in near neutral aqueous solutions.
  • Produces stronger fluorescence at 445 nm in weak alkaline solutions.