Learn More
Medchemexpress LLC 4-Methylumbelliferyl phosphate | 3368-04-5 | 99.8% | 256.15 | 1 ML
SDP
Catalog No. 5000429613
Encompass_Preferred
4-Methylumbelliferyl phosphate (4-MUP) is an anionic organophosphate and a fluorogenic substrate for both acid and alkaline phosphatase. It is also recognized as a nerve agent simulant, primarily for research use.
  • Anionic organophosphate
  • Fluorogenic substrate for acid and alkaline phosphatase
  • Nerve agent simulant
  • Used in serum acid phosphatase assays
Catalog No. 50-004-29613 Supplier Medchemexpress LLC Supplier No. HYD099410MM/1ML
4-Methylumbelliferyl phosphate (4-MUP) is an anionic organophosphate and a fluorogenic substrate for both acid and alkaline phosphatase. It is also recognized as a nerve agent simulant, primarily for research use.
  • Anionic organophosphate
  • Fluorogenic substrate for acid and alkaline phosphatase
  • Nerve agent simulant
  • Used in serum acid phosphatase assays