Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylumbelliferyl phosphate, 99%

Catalog No. AAJ6609106
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6609106 5 g
AAJ66091MD 250 mg
AAJ6609103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6609106 Supplier Thermo Scientific Chemicals Supplier No. J6609106
Only null left
Add to Cart

CAS: 3368-04-5 | C10H9O6P | 256.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3368-04-5
MDL Number MFCD00016969

Specifications

Melting Point 214°C to 216°C
Color White
Molecular Formula C10H9O6P
Quantity 5 g
Beilstein 1687229
Synonym 4-methylumbelliferyl phosphate, 4-methyl-2-oxo-2h-chromen-7-yl dihydrogen phosphate, 4-mup, 2h-1-benzopyran-2-one, 4-methyl-7-phosphonooxy, phosphoric acid 4-methylumbelliferyl ester, 4-methylumbelliferylphosphate, free acid, 4-methyl-2-oxochromen-7-yl oxyphosphonic acid, 5,5-dinitrobarbutyric acid, 4-methylumbelliferone phosphate, 4-methylumbelliferyl phosphate, free acid
Solubility Information Soluble in water
InChI Key BCHIXGBGRHLSBE-UHFFFAOYSA-N
SMILES CC1=CC(=O)OC2=C1C=CC(=C2)OP(=O)(O)O
IUPAC Name (4-methyl-2-oxochromen-7-yl) dihydrogen phosphate
Molecular Weight (g/mol) 256.15
PubChem CID 65118
Formula Weight 256.15
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 4-Methylumbelliferyl phosphate
Show More Show Less
EINECSNumber 222-137-7
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.