Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylumbelliferyl Phosphate 98.0+%, TCI America™
SDP

Catalog No. P1060100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1060100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P1060100MG Supplier TCI America Supplier No. P1060100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3368-04-5
Molecular Formula C10H9O6P
Molecular Weight (g/mol) 256.15
MDL Number MFCD00016969
InChI Key BCHIXGBGRHLSBE-UHFFFAOYSA-N
Synonym 4-methylumbelliferyl phosphate, 4-methyl-2-oxo-2h-chromen-7-yl dihydrogen phosphate, 4-mup, 2h-1-benzopyran-2-one, 4-methyl-7-phosphonooxy, phosphoric acid 4-methylumbelliferyl ester, 4-methylumbelliferylphosphate, free acid, 4-methyl-2-oxochromen-7-yl oxyphosphonic acid, 5,5-dinitrobarbutyric acid, 4-methylumbelliferone phosphate, 4-methylumbelliferyl phosphate, free acid
PubChem CID 65118
IUPAC Name (4-methyl-2-oxochromen-7-yl) dihydrogen phosphate
SMILES CC1=CC(=O)OC2=C1C=CC(=C2)OP(=O)(O)O

Specifications

Melting Point 212°C
Color White
Quantity 100 mg
Formula Weight 256.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylumbelliferyl Phosphate
TSCA Yes
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.