Learn More
Medchemexpress LLC 4-methylumbelliferyl stearate | 79408-85-8 | MFCD00057311 | 99.0% | 442.63 | C28H42O4 | 1 G
SDP
Catalog No. 5000299449
Encompass_Preferred
4-Methylumbelliferyl stearate is a fluorogenic enzymatic substrate that liberates the fluorescent reporter 4-methylumbelliferone upon esterase or lipase-mediated hydrolysis. It is provided as a solid (white to off-white) for use in biochemical assays to quantify enzyme activity and screen inhibitors.
  • Releases highly fluorescent 4-methylumbelliferone upon hydrolysis.
  • Suitable for lipase and esterase activity assays.
  • High purity (≈98.96%) for reproducible results.
  • Solid, white to off-white appearance for easy handling.
  • Long shelf life when stored under recommended conditions.
  • Solubility and handling details available on the technical datasheet.
Catalog No. 50-002-99449 Supplier Medchemexpress LLC Supplier No. HYW0176571G
4-Methylumbelliferyl stearate is a fluorogenic enzymatic substrate that liberates the fluorescent reporter 4-methylumbelliferone upon esterase or lipase-mediated hydrolysis. It is provided as a solid (white to off-white) for use in biochemical assays to quantify enzyme activity and screen inhibitors.
  • Releases highly fluorescent 4-methylumbelliferone upon hydrolysis.
  • Suitable for lipase and esterase activity assays.
  • High purity (≈98.96%) for reproducible results.
  • Solid, white to off-white appearance for easy handling.
  • Long shelf life when stored under recommended conditions.
  • Solubility and handling details available on the technical datasheet.