Learn More
Medchemexpress LLC 4-methylumbelliferyl stearate | 79408-85-8 | MFCD00057311 | 99.0% | 442.63 | C28H42O4 | 500 MG
SDP
Catalog No. 5000300814
Encompass_Preferred
4-methylumbelliferyl stearate is a fluorogenic biochemical reagent used as a substrate to measure lipase and esterase activity. Enzymatic hydrolysis liberates 4-methylumbelliferone, a fluorescent reporter commonly quantified in fluorometric assays. The compound is intended for research use and is provided with supporting COA and SDS documentation.
  • Fluorogenic substrate for lipase and esterase assays.
  • Releases 4-methylumbelliferone upon enzymatic hydrolysis.
  • High purity suitable for quantitative enzymatic measurements.
  • Compatible with common organic solvents for assay preparation.
  • Supplied with COA and SDS for traceability and safety.
Catalog No. 50-003-00814 Supplier Medchemexpress LLC Supplier No. HYW017657500MG
4-methylumbelliferyl stearate is a fluorogenic biochemical reagent used as a substrate to measure lipase and esterase activity. Enzymatic hydrolysis liberates 4-methylumbelliferone, a fluorescent reporter commonly quantified in fluorometric assays. The compound is intended for research use and is provided with supporting COA and SDS documentation.
  • Fluorogenic substrate for lipase and esterase assays.
  • Releases 4-methylumbelliferone upon enzymatic hydrolysis.
  • High purity suitable for quantitative enzymatic measurements.
  • Compatible with common organic solvents for assay preparation.
  • Supplied with COA and SDS for traceability and safety.