Learn More
Medchemexpress LLC 4-methylumbelliferyl stearate | 79408-85-8 | MFCD00057311 | 99.0% | 442.63 g/mol | C28H42O4 | 250 MG
SDP
Catalog No. 5000302586
Encompass_Preferred
4-Methylumbelliferyl stearate is a fluorogenic biochemical reagent used as a substrate for esterase and lipase assays. Enzymatic hydrolysis releases the fluorescent 4-methylumbelliferone (4-MU), enabling sensitive, quantitative measurement of enzyme activity in biochemical and cell-based research. For research use only.
  • Releases fluorescent 4-methylumbelliferone upon hydrolysis.
  • Enables sensitive, quantitative enzyme activity assays.
  • Suitable for esterase and lipase substrate assays.
  • Available in multiple sample sizes for assay optimization.
  • Stable when stored sealed at room temperature.
  • Supplied with purity and batch certificate information.
Catalog No. 50-003-02586 Supplier Medchemexpress LLC Supplier No. 250MGHYW017657
4-Methylumbelliferyl stearate is a fluorogenic biochemical reagent used as a substrate for esterase and lipase assays. Enzymatic hydrolysis releases the fluorescent 4-methylumbelliferone (4-MU), enabling sensitive, quantitative measurement of enzyme activity in biochemical and cell-based research. For research use only.
  • Releases fluorescent 4-methylumbelliferone upon hydrolysis.
  • Enables sensitive, quantitative enzyme activity assays.
  • Suitable for esterase and lipase substrate assays.
  • Available in multiple sample sizes for assay optimization.
  • Stable when stored sealed at room temperature.
  • Supplied with purity and batch certificate information.