Learn More
Selleck Chemical LLC 4-Methylvaleric acid
SDP
Catalog No. 5000003386
Encompass_Preferred
4-Methylvaleric acid (Isocaproic acid Isocaproate 4-Methylpentanoate 4-Methylpentanoic acid) is an important metabolite in early placentas enabling the convertion from cholesterol to pregnenolone to Dehydroepiandrosterone (DHEA)
Catalog No. 50-000-03386 Supplier Selleck Chemical LLC Supplier No. S627825UL
4-Methylvaleric acid (Isocaproic acid Isocaproate 4-Methylpentanoate 4-Methylpentanoic acid) is an important metabolite in early placentas enabling the convertion from cholesterol to pregnenolone to Dehydroepiandrosterone (DHEA)