Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Methylvaleric acid

Catalog No. 2778275G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-782-75G 5 g
27-782-725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 27-782-75G Supplier Sigma Aldrich Supplier No. 2778275G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Conductimetric detection of 4-methylvaleric acid on unfunctionized polymethacrylate resin (tskgel g3000pwxl) columns has been reported.4-methylvaleric acid has been used in asymmetric synthesis of (4s)-4-isopropyl-3-[(2αs)-2α,4α-dimethyl-valeryl)]-2-oxazolidinone.tandem mass spectrometry data independently generated by scripps center for metabolomics is available to view or download in pdf
  • 277827.pdf tested metabolites are featured on scripps center for metabolomics metlin metabolite database
  • To learn more, visit sigma.com/metlin

Specifications

CAS 646-07-1
Density 0.923 g/mL (at 25°C (literature))
Boiling Point 199°C to 201°C (lit.)
Molecular Formula C6H12O2
Refractive Index n20/D 1.414 (literature)
Linear Formula (CH3)2 CHCH2CH2CO2H
MDL Number MFCD00002803
Quantity 5 g
Synonym 4-Methylpentanoic acid; Isocaproic acid
Molecular Weight (g/mol) 116.16
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.