Learn More
Electron Microscopy Sciences Solid Cardboard 4 Microscope Slide Mailer
SDP
Catalog No. 50931176
Encompass_Preferred
Made from .085" (2.17 mm) cardboard. Holds four 3x1" slide.
Catalog No. 50-931-176 Supplier Electron Microscopy Sciences Supplier No. 71541
Made from .085" (2.17 mm) cardboard. Holds four 3x1" slide.