Learn More
HEPATOCHEM 4 ml (1 DRAM) quartz vial
Catalog No. NC3242635
Encompass_Preferred
4 ml (1DRAM) quartz vials 15x45mm with 13-425 thread (cap with septum included)
Catalog No. NC3242635 Supplier HEPATOCHEM Supplier No. HCK102101052
4 ml (1DRAM) quartz vials 15x45mm with 13-425 thread (cap with septum included)