Learn More
Perkin Elmer US LLC 4 mL Clear Glass Screw Top Waste and Wash Vial, 100/pk
Catalog No. 502104674
Encompass_Preferred
Our vials are manufactured from Type I Borosilicate Glass, which meets all USP, JP, and EP Pharmacopeia requirements. The glass performs excellent at high temperatures and is chemical resistant to acidic, neutral and alkali solutions. All our vials are packed in a clean environment to ensure you receive contaminant free product every time.
Catalog No. 50-210-4674 Supplier Perkin Elmer US LLC Supplier No. N9306247
Our vials are manufactured from Type I Borosilicate Glass, which meets all USP, JP, and EP Pharmacopeia requirements. The glass performs excellent at high temperatures and is chemical resistant to acidic, neutral and alkali solutions. All our vials are packed in a clean environment to ensure you receive contaminant free product every time.