Learn More
MESA LABS BOZEMAN MFG FACILITY Mesa Labs 1480 ProSpore Incubator, Incubates 4 mL Biological Indicators, Adjustable Temp 37°C - 60°C
SDP
Catalog No. NC1725835
Encompass
The Mesa Labs Model 1480 ProSpore Incubator is designed for the effective incubation of 4 mL ProSpore biological indicators, crucial for steam sterilization processes. With 11 incubation cavities and configurable temperature settings (37°C, 57°C, 60°C), it ensures precise and efficient operation. Its built-in LED thermometer allows for accurate monitoring, making it a reliable choice in laboratory settings where consistency is key.
This item is not returnable. View return policy
Catalog No. NC1725835 Supplier MESA LABS BOZEMAN MFG FACILITY Supplier No. 7706I1480M
The Mesa Labs Model 1480 ProSpore Incubator is designed for the effective incubation of 4 mL ProSpore biological indicators, crucial for steam sterilization processes. With 11 incubation cavities and configurable temperature settings (37°C, 57°C, 60°C), it ensures precise and efficient operation. Its built-in LED thermometer allows for accurate monitoring, making it a reliable choice in laboratory settings where consistency is key.