Learn More
Medchemexpress LLC HY-118480 5mg Medchemexpress, 4-MMPB CAS:928853-86-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1184805MG
Encompass_Preferred
Medchemexpress, HY-118480 5mg 4-MMPB CAS:928853-86-5 4-MMPB is a selective inhibitor of 15-lipoxygenase, with an IC50 of 18 μM. 4-MMPB has IC50s of 19.5 μM and 19.1 μM for soybean 15-lipoxygenase (SLO) and human 15-lipoxygenase-1 (15-LOX-1), respectively. 4-MMPB has potential for the research of prostate cancer[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0915