Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-MMPB | 928853-86-5 | 313.42 g/mol | C16H19N5S | 25 MG
SDP

Catalog No. 5000227345
Encompass_Preferred

4-MMPB is a small-molecule research compound that acts as a selective 15-lipoxygenase inhibitor and is provided for laboratory research use only. It has documented enzyme inhibition and cell viability data and is commonly used in biochemical assays and exploratory cellular studies.

  • Selective 15-lipoxygenase inhibition (IC50 ≈ 18 μM)
  • Molecular formula C16H19N5S; molecular weight 313.42 g/mol
  • Soluble in DMSO (7.69 mg/mL; may require sonication)
  • Compatible in vivo formulations at ≥ 0.77 mg/mL in multiple vehicle systems
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years)
  • Intended for laboratory research use only; not for human or clinical use

Catalog No. 50-002-27345 Supplier Medchemexpress LLC Supplier No. HY11848025MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-MMPB is a small-molecule research compound that acts as a selective 15-lipoxygenase inhibitor and is provided for laboratory research use only. It has documented enzyme inhibition and cell viability data and is commonly used in biochemical assays and exploratory cellular studies.

  • Selective 15-lipoxygenase inhibition (IC50 ≈ 18 μM)
  • Molecular formula C16H19N5S; molecular weight 313.42 g/mol
  • Soluble in DMSO (7.69 mg/mL; may require sonication)
  • Compatible in vivo formulations at ≥ 0.77 mg/mL in multiple vehicle systems
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years)
  • Intended for laboratory research use only; not for human or clinical use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.