Learn More
Medchemexpress LLC 4-mmpb | 928853-86-5 | 98.5% | 313.42 | C16H19N5S | 10 MG
SDP
Catalog No. 5000307679
Encompass_Preferred
4-MMPB is a research-grade small molecule that selectively inhibits 15-lipoxygenase and is provided for biochemical and cell-based studies of lipid metabolism and cancer-related pathways.
  • Selective inhibitor of 15-lipoxygenase with a reported IC50 of ≈18 μM.
  • High reported purity, suitable for biochemical assays (≈98.5%).
  • Molecular formula C16H19N5S and molecular weight 313.42 g/mol.
  • Supplied as a solid 10 MG and available in larger quantities; also offered as a 10 mM solution in DMSO.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-003-07679 Supplier Medchemexpress LLC Supplier No. HY11848010MG
4-MMPB is a research-grade small molecule that selectively inhibits 15-lipoxygenase and is provided for biochemical and cell-based studies of lipid metabolism and cancer-related pathways.
  • Selective inhibitor of 15-lipoxygenase with a reported IC50 of ≈18 μM.
  • High reported purity, suitable for biochemical assays (≈98.5%).
  • Molecular formula C16H19N5S and molecular weight 313.42 g/mol.
  • Supplied as a solid 10 MG and available in larger quantities; also offered as a 10 mM solution in DMSO.
  • Intended for research use only; not for human or clinical use.