Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-MMPB | 928853-86-5 | 98.5% | 313.42 g/mol | C16H19N5S | 50 MG
SDP

Catalog No. 5000226824
Encompass_Preferred

4-MMPB is a research reagent and selective 15-lipoxygenase inhibitor used in biochemical and pharmacological studies. Supplied as a solid with characterization data and supporting documents (COA, SDS), it is intended for research use only.

  • Solid, light yellow to orange appearance
  • Purity 98.5% (manufacturer reported)
  • Molecular weight 313.42 g/mol, formula C16H19N5S
  • Available in small milligram packages including 50 MG
  • Includes datasheet, certificate of analysis, and SDS for characterization and safety
  • Store powder at -20°C (3 years) or 4°C (2 years); in solvent store at -80°C or -20°C

Catalog No. 50-002-26824 Supplier Medchemexpress LLC Supplier No. HY11848050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-MMPB is a research reagent and selective 15-lipoxygenase inhibitor used in biochemical and pharmacological studies. Supplied as a solid with characterization data and supporting documents (COA, SDS), it is intended for research use only.

  • Solid, light yellow to orange appearance
  • Purity 98.5% (manufacturer reported)
  • Molecular weight 313.42 g/mol, formula C16H19N5S
  • Available in small milligram packages including 50 MG
  • Includes datasheet, certificate of analysis, and SDS for characterization and safety
  • Store powder at -20°C (3 years) or 4°C (2 years); in solvent store at -80°C or -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.