Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Morpholine-4-sulfonyl)-benzenesulfonyl chloride, Technical Grade, Thermo Scientific™

Catalog No. MO00516DA
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO00516DA 1 g Amber Glass Bottle
MO00516CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MO00516DA Supplier Thermo Scientific Chemicals Supplier No. MO00516DA
Add to Cart

Chemical Identifiers

CAS 465514-13-0
Molecular Formula C10H12ClNO5S2
Molecular Weight (g/mol) 325.778
InChI Key ZZSBUDUHESWZJB-UHFFFAOYSA-N
Synonym 4-morpholin-4-ylsulfonyl benzenesulfonyl chloride, 4-morpholine-4-sulfonyl benzene-1-sulfonyl chloride, 4-morpholinosulfonyl benzene-1-sulfonyl chloride, 4-morpholine-4-sulfonyl benzenesulfonyl chloride, 4-morpholine-4-sulfonyl-benzenesulfonyl chloride, 4-morpholine-4-sulfonyl phenylsulfonyl chloride, 4-morpholine-4-sulfonyl-benzenesulfonylchloride
PubChem CID 2779790
IUPAC Name 4-morpholin-4-ylsulfonylbenzenesulfonyl chloride
SMILES C1COCCN1S(=O)(=O)C2=CC=C(C=C2)S(=O)(=O)Cl

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 325.79
Percent Purity Tech.
Grade Technical
Chemical Name or Material 4-(Morpholine-4-sulfonyl)-benzenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.