Learn More
Medchemexpress LLC 4-Morpholineethanesulfonic acid-13d | 352534-94-2 | 98.8% | 100 MG
SDP
Catalog No. 5000409964
Encompass_Preferred
MES-d13 is the deuterium labeled MES, an amphoteric ion buffer with a buffer capacity ranging from pH 5.5-7.0. It serves as a Good's buffer, widely utilized in various biochemistry and molecular biology experiments. This compound can be used as a tracer and as an internal standard for quantitative analysis.
  • Widely used in cell culture, enzyme activity determination, electrophoresis, and protein studies.
  • Can be used as a tracer for quantitation during drug development.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-09964 Supplier Medchemexpress LLC Supplier No. HYD0858S100MG
MES-d13 is the deuterium labeled MES, an amphoteric ion buffer with a buffer capacity ranging from pH 5.5-7.0. It serves as a Good's buffer, widely utilized in various biochemistry and molecular biology experiments. This compound can be used as a tracer and as an internal standard for quantitative analysis.
  • Widely used in cell culture, enzyme activity determination, electrophoresis, and protein studies.
  • Can be used as a tracer for quantitation during drug development.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.